Assay Details
Gene Symbol: SPG11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003640025.4 | XP_003640073.2 | 37 - 38 | 6858 | 161 | |
XM_005638363.3 | XP_005638420.1 | 34 - 35 | 6106 | 161 | ||
XM_014109490.2 | XP_013964965.1 | 36 - 37 | 6654 | 161 | ||
XM_014109491.2 | XP_013964966.1 | 36 - 37 | 6633 | 161 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
