Assay Details
Gene Symbol: FILIP1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005639388.3 | XP_005639445.1 | 3730 | 120 | ||
XM_022413927.1 | XP_022269635.1 | 3382 | 120 | |||
XM_022413928.1 | XP_022269636.1 | 3159 | 120 | |||
XM_022413929.1 | XP_022269637.1 | 3154 | 120 | |||
XM_022413930.1 | XP_022269638.1 | 3034 | 120 | |||
XM_022413931.1 | XP_022269639.1 | 3068 | 120 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() |
