Assay Details
Gene Symbol: WDR20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022422389.1 | XP_022278097.1 | 3 | 478 | 95 | |
XM_022422390.1 | XP_022278098.1 | 3 | 531 | 95 | ||
XM_022422391.1 | XP_022278099.1 | 3 | 341 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
