Assay Details
Gene Symbol: MYCL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005628887.3 | XP_005628944.1 | 6 | 1620 | 74 | |
XM_005628888.3 | XP_005628945.1 | 3 | 932 | 74 | ||
XM_014119331.2 | XP_013974806.1 | 8 | 2168 | 74 | ||
XM_014119333.2 | XP_013974808.1 | 7 | 2643 | 74 | ||
XM_022427767.1 | XP_022283475.1 | 6 | 1661 | 74 | ||
XM_022427768.1 | XP_022283476.1 | 8 | 2165 | 74 | ||
XM_022427769.1 | XP_022283477.1 | 6 | 1634 | 74 | ||
XM_022427771.1 | XP_022283479.1 | 7 | 2647 | 74 | ||
XM_022427772.1 | XP_022283480.1 | 6 | 1624 | 74 | ||
XM_022427773.1 | XP_022283481.1 | 6 | 1545 | 74 | ||
XM_022427774.1 | XP_022283482.1 | 5 | 1227 | 74 | ||
XM_022427775.1 | XP_022283483.1 | 5 | 1196 | 74 | ||
XM_022427776.1 | XP_022283484.1 | 5 | 1107 | 74 | ||
XM_022427777.1 | XP_022283485.1 | 5 | 1182 | 74 | ||
XM_022427778.1 | XP_022283486.1 | 4 | 1113 | 74 | ||
XM_022427779.1 | XP_022283487.1 | 5 | 2091 | 74 | ||
XM_022427780.1 | XP_022283488.1 | 4 | 1068 | 74 | ||
XM_022427781.1 | XP_022283489.1 | 3 | 1012 | 74 | ||
XM_539578.6 | XP_539578.2 | 3 | 949 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
