Assay Details
Gene Symbol: BDKRB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003095.1 | NP_001003095.1 | 3 | 703 | 103 | |
XM_005623175.3 | XP_005623232.1 | 4 | 1284 | 103 | ||
GenBank mRNA | AF334948.1 | - | 3 | 703 | 103 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
