Assay Details
Gene Symbol: PDE6G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003235.1 | NP_001003235.1 | 383 | 87 | ||
XM_022422536.1 | XP_022278244.1 | 1105 | 87 | |||
GenBank mRNA | U49359.1 | - | 383 | 87 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
