Assay Details
Gene Symbol: ELAVL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005632851.3 | XP_005632908.1 | 3 - 4 | 739 | 98 | |
XM_005632852.3 | XP_005632909.1 | 3 - 4 | 739 | 98 | ||
XM_022407022.1 | XP_022262730.1 | 3 - 4 | 736 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
