Assay Details
Gene Symbol: RNASEL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001097549.1 | NP_001091018.1 | 692 | 170 | ||
XM_005622259.3 | XP_005622316.1 | 629 | 170 | |||
XM_014114972.2 | XP_013970447.1 | 698 | 170 | |||
XM_014114973.2 | XP_013970448.1 | 690 | 170 | |||
XM_014114974.2 | XP_013970449.1 | 684 | 170 | |||
XM_022420760.1 | XP_022276468.1 | 684 | 170 | |||
GenBank mRNA | DQ497163.1 | - | 692 | 170 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
