Assay Details
Gene Symbol: BRCA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006653.4 | NP_001006654.2 | 8603 | 148 | ||
XM_014107324.2 | XP_013962799.1 | 8920 | 148 | |||
XM_022409552.1 | XP_022265260.1 | 8543 | 148 | |||
XM_022409553.1 | XP_022265261.1 | 8408 | 148 | |||
GenBank mRNA | AB043895.5 | - | 8603 | 148 | ||
AB622997.1 | - | 8392 | 148 | |||
AB622998.1 | - | 8392 | 148 | |||
AB622999.1 | - | 8392 | 148 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() |
