Assay Details
Gene Symbol: CHRNB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022421343.1 | XP_022277051.1 | 2 - 3 | 470 | 149 | |
XM_547565.5 | XP_547565.2 | 2 - 3 | 1391 | 149 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_022421343.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
