Assay Details
Gene Symbol: HPS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005637599.3 | XP_005637656.1 | 13 - 14 | 1866 | 115 | |
XM_005637600.3 | XP_005637657.1 | 13 - 14 | 1513 | 115 | ||
XM_005637601.2 | XP_005637658.1 | 13 - 14 | 1870 | 115 | ||
XM_022411759.1 | XP_022267467.1 | 12 - 13 | 1701 | 115 | ||
XM_022411760.1 | XP_022267468.1 | 4 - 5 | 570 | 115 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() |
