Assay Details
Gene Symbol: NCK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005625984.3 | XP_005626041.1 | 4 - 5 | 951 | 170 | |
XM_005625985.3 | XP_005626042.1 | 4 - 5 | 663 | 170 | ||
XM_005625986.3 | XP_005626043.1 | 4 - 5 | 956 | 170 | ||
XM_014117410.2 | XP_013972885.1 | 3 - 4 | 592 | 170 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
