Assay Details
Gene Symbol: MLLT10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022406741.1 | XP_022262449.1 | 272 | 134 | ||
XM_022406745.1 | XP_022262453.1 | 272 | 134 | |||
XM_022406750.1 | XP_022262458.1 | 769 | 134 | |||
XM_022406753.1 | XP_022262461.1 | 649 | 134 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_022406750.1, XM_022406753.1 transcripts.
Set Membership: |
![]() ![]() |
