Assay Details
Gene Symbol: MLLT10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022406741.1 | XP_022262449.1 | 1482 | 144 | ||
XM_022406745.1 | XP_022262453.1 | 1482 | 144 | |||
XM_022406750.1 | XP_022262458.1 | 1979 | 144 | |||
XM_022406753.1 | XP_022262461.1 | 1859 | 144 | |||
XM_022406756.1 | XP_022262464.1 | 861 | 144 |

More Information
Set Membership: |
![]() ![]() |
