Assay Details
Gene Symbol: CCDC189 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195695.1 | NP_001182624.1 | 776 | 120 | ||
XM_005621225.3 | XP_005621282.1 | 870 | 120 | |||
XM_022419489.1 | XP_022275197.1 | 870 | 120 | |||
XM_022419490.1 | XP_022275198.1 | 597 | 120 | |||
GenBank mRNA | DN748234.1 | - | 309 | 120 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() |
