Assay Details
Gene Symbol: MYZAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005638457.1 | XP_005638514.1 | 9 - 10 | 1022 | 174 | |
XM_022412856.1 | XP_022268564.1 | 9 - 10 | 1022 | 174 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() |
