Assay Details
Gene Symbol: CCNO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_544336.5 | XP_544336.2 | 2 - 3 | 814 | 142 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: CCNO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_544336.5 | XP_544336.2 | 2 - 3 | 814 | 142 |
Set Membership: |
![]() ![]() ![]() ![]() |