Assay Details
Gene Symbol: ETV6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005637150.3 | XP_005637207.1 | 6 - 7 | 1204 | 117 | |
XM_005637152.3 | XP_005637209.1 | 5 - 6 | 1058 | 117 | ||
XM_022411287.1 | XP_022266995.1 | 6 - 7 | 1526 | 117 | ||
XM_543812.6 | XP_543812.3 | 6 - 7 | 1207 | 117 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
