Assay Details
Gene Symbol: LIF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001197073.1 | NP_001184002.1 | 2 - 3 | 354 | 135 | |
XM_005636000.2 | XP_005636057.1 | 3 - 4 | 2396 | 135 | ||
XM_022410147.1 | XP_022265855.1 | 4 - 5 | 1312 | 135 | ||
GenBank mRNA | AF512028.1 | - | 1 - 2 | 138 | 135 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
