Assay Details
Gene Symbol: BABAM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005630287.2 | XP_005630344.1 | 9 - 10 | 1020 | 158 | |
XM_022404584.1 | XP_022260292.1 | 9 - 10 | 1004 | 158 | ||
XM_022404585.1 | XP_022260293.1 | 9 - 10 | 1020 | 158 | ||
XM_845588.4 | XP_850681.1 | 9 - 10 | 1020 | 158 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() |
