Assay Details
Gene Symbol: CEMIP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005615860.3 | XP_005615917.1 | 16 - 17 | 2972 | 115 | |
XM_022421202.1 | XP_022276910.1 | 15 - 16 | 2770 | 115 | ||
XM_541287.6 | XP_541287.3 | 15 - 16 | 2702 | 115 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
