Assay Details
Gene Symbol: LOC100855783 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014107689.2 | XP_013963164.1 | 29 - 30 | 3520 | 130 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC100855783 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014107689.2 | XP_013963164.1 | 29 - 30 | 3520 | 130 |
Set Membership: |
![]() ![]() ![]() ![]() |