Assay Details
Gene Symbol: LPAR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014121767.2 | XP_013977242.1 | 3 | 1010 | 166 | |
XM_022406813.1 | XP_022262521.1 | 4 | 1113 | 166 | ||
XM_022406814.1 | XP_022262522.1 | 4 | 1099 | 166 | ||
XM_022406815.1 | XP_022262523.1 | 4 | 1045 | 166 | ||
XM_022406816.1 | XP_022262524.1 | 4 | 1138 | 166 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
