Assay Details
Gene Symbol: BMI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001287063.1 | NP_001273992.1 | 718 | 96 | ||
XM_005617014.3 | XP_005617071.1 | 693 | 96 | |||
XM_005617015.3 | XP_005617072.1 | 635 | 96 | |||
XM_022405588.1 | XP_022261296.1 | 512 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
