Assay Details
Gene Symbol: CRK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003435202.4 | XP_003435250.1 | 2 | 515 | 105 | |
XM_003435203.4 | XP_003435251.1 | 2 | 515 | 105 | ||
XM_022423445.1 | XP_022279153.1 | 2 | 518 | 105 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
