Assay Details
Gene Symbol: PURA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005617272.3 | XP_005617329.1 | 1504 | 118 | ||
XM_005617273.3 | XP_005617330.1 | 1365 | 118 | |||
XM_005617274.3 | XP_005617331.1 | 1223 | 118 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() |
