Assay Details
Gene Symbol: IRAG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005637049.3 | XP_005637106.1 | 15 - 16 | 1373 | 77 | |
XM_005637050.3 | XP_005637107.1 | 15 - 16 | 1370 | 77 | ||
XM_005637051.3 | XP_005637108.1 | 14 - 15 | 1265 | 77 | ||
XM_543758.6 | XP_543758.2 | 14 - 15 | 1503 | 77 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() |
