Assay Details
Gene Symbol: NCK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005634467.3 | XP_005634524.1 | 4 - 5 | 1205 | 121 | |
XM_005634468.2 | XP_005634525.1 | 3 - 4 | 1054 | 121 | ||
XM_005634470.3 | XP_005634527.1 | 3 - 4 | 1052 | 121 | ||
XM_005634471.2 | XP_005634528.1 | 2 - 3 | 830 | 121 | ||
XM_846945.5 | XP_852038.1 | 3 - 4 | 1085 | 121 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
