Assay Details
Gene Symbol: GLRA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014107468.2 | XP_013962943.1 | 8 - 9 | 1554 | 157 | |
XM_014107469.2 | XP_013962944.1 | 9 - 10 | 1165 | 157 | ||
XM_022409916.1 | XP_022265624.1 | 9 - 10 | 1378 | 157 | ||
XM_022409917.1 | XP_022265625.1 | 8 - 9 | 1054 | 157 | ||
XM_022409918.1 | XP_022265626.1 | 8 - 9 | 1262 | 157 | ||
XM_022409919.1 | XP_022265627.1 | 8 - 9 | 1553 | 157 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
