Assay Details
Gene Symbol: PKIA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003433669.4 | XP_003433717.1 | 3 | 366 | 133 | |
XM_005638060.3 | XP_005638117.2 | 2 | 136 | 133 | ||
XM_005638062.2 | XP_005638119.1 | 3 | 289 | 133 | ||
XM_022412474.1 | XP_022268182.1 | 2 | 232 | 133 | ||
XM_848290.4 | XP_853383.2 | 2 | 226 | 133 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() |
