Assay Details
Gene Symbol: ARHGAP45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022407306.1 | XP_022263014.1 | 11 - 12 | 1310 | 85 | |
XM_022407307.1 | XP_022263015.1 | 11 - 12 | 1318 | 85 | ||
XM_022407308.1 | XP_022263016.1 | 10 - 11 | 1309 | 85 | ||
XM_022407309.1 | XP_022263017.1 | 10 - 11 | 1307 | 85 | ||
XM_022407310.1 | XP_022263018.1 | 10 - 11 | 1308 | 85 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() |
