Assay Details
Gene Symbol: MAP3K20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022414748.1 | XP_022270456.1 | 6 - 7 | 613 | 137 | |
XM_022414749.1 | XP_022270457.1 | 6 - 7 | 721 | 137 | ||
XM_022414750.1 | XP_022270458.1 | 6 - 7 | 613 | 137 | ||
XM_535966.6 | XP_535966.2 | 7 - 8 | 864 | 137 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() |
