Assay Details
Gene Symbol: INTS13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001197314.1 | NP_001184243.1 | 13 | 1777 | 147 | |
XM_005636713.3 | XP_005636770.1 | 14 | 2329 | 147 | ||
XM_005636714.3 | XP_005636771.1 | 14 | 1915 | 147 | ||
XM_022410862.1 | XP_022266570.1 | 12 | 2149 | 147 | ||
XM_022410863.1 | XP_022266571.1 | 13 | 2093 | 147 | ||
XM_022410864.1 | XP_022266572.1 | 12 | 2018 | 147 | ||
GenBank mRNA | GU811149.1 | - | 13 | 1777 | 147 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
