Assay Details
Gene Symbol: PPP4R3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005626116.3 | XP_005626173.1 | 5 - 6 | 1229 | 148 | |
XM_005626117.3 | XP_005626174.1 | 5 - 6 | 1231 | 148 | ||
XM_022424761.1 | XP_022280469.1 | 5 - 6 | 1227 | 148 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
