Assay Details
Gene Symbol: BEGAIN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005623810.2 | XP_005623867.1 | 5 - 6 | 443 | 141 | |
XM_005623811.3 | XP_005623868.1 | 5 - 6 | 460 | 141 | ||
XM_005623812.3 | XP_005623869.1 | 5 - 6 | 478 | 141 | ||
XM_005623813.2 | XP_005623870.1 | 4 - 5 | 353 | 141 | ||
XM_005623814.3 | XP_005623871.1 | 4 - 5 | 370 | 141 | ||
XM_005623815.3 | XP_005623872.1 | 4 - 5 | 387 | 141 | ||
XM_005623816.3 | XP_005623873.1 | 4 - 5 | 657 | 141 | ||
XM_022422375.1 | XP_022278083.1 | 5 - 6 | 751 | 141 | ||
XM_022422377.1 | XP_022278085.1 | 3 - 4 | 203 | 141 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
