Assay Details
Gene Symbol: IL22RA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022413330.1 | XP_022269038.1 | 5 - 6 | 752 | 190 | |
XM_850020.4 | XP_855113.2 | 6 - 7 | 885 | 190 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
