Assay Details
Gene Symbol: RSKR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005624861.3 | XP_005624918.1 | 3 - 4 | 431 | 151 | |
XM_014116780.2 | XP_013972255.1 | 3 - 4 | 427 | 151 |

More Information
Set Membership: |
![]() ![]() ![]() |
