Assay Details
Gene Symbol: ELAPOR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014118966.2 | XP_013974441.1 | 8 - 9 | 1011 | 98 | |
XM_022427285.1 | XP_022282993.1 | 7 - 8 | 883 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
