Assay Details
Gene Symbol: NMNAT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005634506.3 | XP_005634563.1 | 6 | 625 | 149 | |
XM_005634510.3 | XP_005634567.1 | 5 | 695 | 149 | ||
XM_014106717.2 | XP_013962192.1 | 7 | 736 | 149 | ||
XM_014106718.2 | XP_013962193.1 | 7 | 880 | 149 | ||
XM_014106719.2 | XP_013962194.1 | 7 | 731 | 149 | ||
XM_014106721.2 | XP_013962196.1 | 5 | 589 | 149 | ||
XM_022408690.1 | XP_022264398.1 | 6 | 700 | 149 | ||
XM_022408691.1 | XP_022264399.1 | 7 | 882 | 149 | ||
XM_022408692.1 | XP_022264400.1 | 7 | 761 | 149 | ||
XM_022408693.1 | XP_022264401.1 | 7 | 666 | 149 | ||
XM_022408694.1 | XP_022264402.1 | 3 | 322 | 149 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
