Assay Details
Gene Symbol: ZNF644 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005621941.3 | XP_005621998.1 | 5 | 3135 | 111 | |
XM_005621943.3 | XP_005622000.1 | 4 | 2992 | 111 | ||
XM_014114846.2 | XP_013970321.1 | 3 | 2347 | 111 | ||
XM_022420510.1 | XP_022276218.1 | 5 | 3286 | 111 | ||
XM_022420511.1 | XP_022276219.1 | 3 | 2931 | 111 | ||
XM_022420512.1 | XP_022276220.1 | 2 | 2286 | 111 | ||
XM_022420513.1 | XP_022276221.1 | 3 | 2347 | 111 | ||
XM_022420514.1 | XP_022276222.1 | 3 | 2347 | 111 | ||
XM_848131.5 | XP_853224.2 | 3 | 2347 | 111 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
