Assay Details
Gene Symbol: IQSEC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005637326.3 | XP_005637383.1 | 9 - 10 | 2917 | 110 | |
XM_014108641.2 | XP_013964116.1 | 9 - 10 | 2917 | 110 | ||
XM_014108642.2 | XP_013964117.1 | 9 - 10 | 2917 | 110 | ||
XM_014108643.2 | XP_013964118.1 | 9 - 10 | 2917 | 110 | ||
XM_543874.5 | XP_543874.2 | 9 - 10 | 2918 | 110 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
