Assay Details
Gene Symbol: BBX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005639459.2 | XP_005639516.1 | 6 - 7 | 898 | 110 | |
XM_005639460.3 | XP_005639517.1 | 6 - 7 | 847 | 110 | ||
XM_005639461.2 | XP_005639518.1 | 7 - 8 | 1016 | 110 | ||
XM_005639462.3 | XP_005639519.1 | 4 - 5 | 799 | 110 | ||
XM_005639463.2 | XP_005639520.1 | 6 - 7 | 898 | 110 | ||
XM_005639464.2 | XP_005639521.1 | 6 - 7 | 898 | 110 | ||
XM_005639465.2 | XP_005639522.1 | 6 - 7 | 898 | 110 | ||
XM_005639466.2 | XP_005639523.1 | 6 - 7 | 898 | 110 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
