Assay Details
Gene Symbol: GABRB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014113039.2 | XP_013968514.1 | 1 - 2 | 83 | 102 | |
XM_014113040.2 | XP_013968515.1 | 1 - 2 | 83 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
