Assay Details
Gene Symbol: NPFFR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_846842.3 | XP_851935.2 | 3 - 4 | 643 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
