Assay Details
Gene Symbol: TENT4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005617576.2 | XP_005617633.1 | 6 - 7 | 1160 | 163 | |
XM_005617577.2 | XP_005617634.1 | 6 - 7 | 1169 | 163 | ||
XM_022411501.1 | XP_022267209.1 | 6 - 7 | 1163 | 163 | ||
XM_535307.6 | XP_535307.5 | 6 - 7 | 1163 | 163 |

More Information
Set Membership: |
![]() ![]() ![]() |
