Assay Details
Gene Symbol: AGPAT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005627177.3 | XP_005627234.1 | 3 - 4 | 639 | 147 | |
XM_005627178.3 | XP_005627235.1 | 3 - 4 | 598 | 147 | ||
XM_005627180.3 | XP_005627237.1 | 3 - 4 | 500 | 147 | ||
XM_022425868.1 | XP_022281576.1 | 4 - 5 | 1474 | 147 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() |
