Assay Details
Gene Symbol: FGF7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003237.1 | NP_001003237.1 | 2 - 3 | 469 | 162 | |
XM_005638175.3 | XP_005638232.1 | 3 - 4 | 794 | 162 | ||
GenBank mRNA | U80800.1 | - | 2 - 3 | 469 | 162 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
