Assay Details
Gene Symbol: RNF135 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005624827.3 | XP_005624884.1 | 5 | 1291 | 154 | |
XM_022423317.1 | XP_022279025.1 | 5 | 803 | 154 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
