Assay Details
Gene Symbol: HPSE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022411603.1 | XP_022267311.1 | 7 - 8 | 1211 | 85 | |
XM_845218.5 | XP_850311.1 | 7 - 8 | 1209 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
