Assay Details
Gene Symbol: ZBTB18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_547497.6 | XP_547497.2 | 1 - 2 | 1214 | 138 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_547497.6 transcripts.
Set Membership: |
![]() ![]() ![]() |
